missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-2-carboxamide, 98%

Product Code. 11348135
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348135 10 g 10g
11338135 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348135 Supplier Thermo Scientific Alfa Aesar Supplier No. L08391.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2243-82-5 | C11H9NO | 171.199 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2243-82-5
Molecular Formula C11H9NO
Molecular Weight (g/mol) 171.199
MDL Number MFCD00016812
InChI Key JVXXKQIRGQDWOJ-UHFFFAOYSA-N
Synonym 2-naphthamide, 2-naphthalenecarboxamide, naphthalene-2-carboxylic acid amide, 2naphthylamide, beta-naphthamide, beta-naphthylamide, isonaphthoic amide, .beta.-naphthamid, 2-naphthamide #, .beta.-naphthamide
PubChem CID 75245
IUPAC Name naphthalene-2-carboxamide
SMILES C1=CC=C2C=C(C=CC2=C1)C(=O)N

Specifications

Melting Point 193°C to 197°C
Quantity 10 g
Beilstein 1101108
Formula Weight 171.2
Percent Purity 98%
Chemical Name or Material Naphthalene-2-carboxamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.