Learn More
Naphthalene, 99%
Description
Naphthalene is formerly important in dyestuff manufacture. It acts as a precursor to phthalic anhydride. Naphthalene is used in the manufacturing of phthalic anhydride, naphthalene sulfonic acids and hydrogenated naphthalenes such as 1,2,3,4-tetrahydronaphthalene, which is used as a precursor for various dyestuffs, pigments, rubber processing chemicals and in pharmaceuticals. Its molten form is used as solvent replacing the high- boiling solvents like dichlorobenzene, nitrobenzene and durene. It is involved as reaction medium for in the Diels-Alder reaction between C60 with anthracene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 80°C to 82°C |
| Density | 0.9628 g/mL |
| Boiling Point | 218°C |
| Flash Point | 78°C (172°F) |
| Odor | Aromatic |
| Refractive Index | 1.5821 |
| Quantity | 2 kg |
| UN Number | UN1334 |
| Beilstein | 1421310 |
| Merck Index | 14,6370 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.