accessibility menu, dialog, popup

UserName

CAS RN 91-20-3

IUPAC Name: naphthalene
Synonyms: naphthalene naftalen naftaleno caswell no. 587 epa pesticide chemical code 055801 camphor tar dezodorator naphtalene naphtaline naphthalin
Molecular Weight (g/mol): 128.17
Molecular Formula: C10H8
InChi Key: UFWIBTONFRDIAS-UHFFFAOYSA-N
SMILES: C1=CC2=CC=CC=C2C=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (6)
  • (4)
  • (3)
Boiling Point 
  • (7)
  • (4)
Physical Form 
  • (3)
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (6)
  • (4)
  • (3)
  • (7)
  • (4)
  • (3)
  • (4)
  • (1)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (3)