missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene, 99+%, scintillation grade

Código de producto. 10417960 Tienda Thermo Scientific Chemicals Productos
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass Bottle
Tamaño de la unidad:
1kg
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Packaging unitSize
10417960 5 g Glass Bottle 5g
10792601 250 g Glass Bottle 250g
10124390 1 kg Glass Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10417960 Proveedor Thermo Scientific Acros N.º de proveedor 180200050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 91-20-3 | C10H8 | 128.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For fluorescence/luminescence spectroscopy
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 91-20-3
Molecular Formula C10H8
Molecular Weight (g/mol) 128.17
MDL Number MFCD00001742
InChI Key UFWIBTONFRDIAS-UHFFFAOYSA-N
PubChem CID 931
ChEBI CHEBI:16482
IUPAC Name naphthalene
SMILES C1=CC2=CC=CC=C2C=C1

Especificaciones

Boiling Point 218.0°C
Melting Point 79.0°C to 82.0°C
Physical Form Powder
CAS Min % 99.0
CAS Max % 100.0
Density 0.9900g/mL
Color Beige to Red/Salmon
Quantity 5 g
Flash Point 78°C
Assay Percent Range 99+%
Infrared Spectrum Authentic
Linear Formula C10H8
Beilstein 05, IV, 1640
Merck Index 15, 6455
Packaging Glass Bottle
Solubility Information Solubility in water: 30 mg/l (25°C). Other solubilities: soluble in ethanol,, very soluble in acetone, chloroform, ether,, benzene, toluene, carbon disulfide
Formula Weight 128.17
Percent Purity ≥99%
Specific Gravity 0.99
Chemical Name or Material Naphthalene, 99+%
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Naphthalene
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Flammable solid Category 2
Indicaciones de peligro
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H351-Suspected of causing cancer.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Consejos de prudencia
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
Otros peligros
  • MIXTURE LIST-Contain: Naphthalene

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.