missing translation for 'onlineSavingsMsg'
Learn More

α-Naphtholbenzion, MP Biomedicals™

Product Code. 11424981 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424981 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11424981 Supplier MP Biomedicals Supplier No. 0215578525
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 6948-88-5
Molecular Formula C27H20O3
Molecular Weight (g/mol) 392.454
InChI Key OUYLDJXFDLBCTQ-UHFFFAOYSA-N
Synonym bis 4-hydroxy-1-naphthyl phenylmethanol, 4,4'-hydroxy phenyl methylene bis naphthalen-1-ol, 4-hydroxy-alpha-4-hydroxynaphthyl-alpha-phenylnaphthalene-1-methanol, 4-hydroxy-4-hydroxynaphthalen-1-yl-phenyl-methyl naphthalen-1-ol, bis 4-hydroxynaphthyl phenylmethan-1-ol, 4,4'-hydroxy phenyl methylene di 1-naphthol, 4,4-alpha-hydroxybenzylidene di-1-naphthol, bis-4-hydroxy-1 naphthyl-phenyl-methanol, 4,4-hydroxy phenyl methylene bis naphthalen-1-ol, 4-hydroxy 4-hydroxynaphthalen-1-yl benzyl naphthalen-1-ol
PubChem CID 81382
IUPAC Name 4-[hydroxy-(4-hydroxynaphthalen-1-yl)-phenylmethyl]naphthalen-1-ol
SMILES C1=CC=C(C=C1)C(C2=CC=C(C3=CC=CC=C32)O)(C4=CC=C(C5=CC=CC=C54)O)O

Specifications

Color Red to Brown
Quantity 25 g
Formula Weight 392.5
Physical Form Powder
Chemical Name or Material α-Naphtholbenzion
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.