missing translation for 'onlineSavingsMsg'
Learn More

Naproxen sodium, MedChemExpress

Product Code. 16436298
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16436298 10 g 10g
16428887 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16436298 Supplier Medchem Express Supplier No. HY15030A
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Naproxen sodium is a COX-1 and COX-2 inhibitor with IC50s of 8.72 and 5.15 μM, respectively in cell assay.

Naproxen sodium is a COX-1 and COX-2 inhibitor with IC50s of 8.72 and 5.15 μM, respectively in cell assay.

Specifications

Chemical Name or Material Naproxen sodium
CAS 26159-34-2
Color White
Purity Grade Notes Research
Molecular Formula C14H13NaO3
Quantity 10 g
Solubility Information H2O : 75 mg/mL (297.34 mM; Need ultrasonic and warming) ∣DMSO : 5 mg/mL (19.82 mM; Need ultrasonic)
SMILES C[C@H](C(O[Na])=O)C1=CC2=CC=C(OC)C=C2C=C1
Molecular Weight (g/mol) 252.24
Formula Weight 252.24
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.