Learn More
NAPRT1 Antibody (CL0366) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52924MFV610
Product Code. 30302240
  • 780.00 € / 0.10mL
Description
NAPRT1 Monoclonal antibody specifically detects NAPRT1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NAPRT1
Monoclonal
mFluor Violet 610 SE
EC 2.4.2.11, FHA-HIT-interacting protein, FHIP, NAPRTase, nicotinate phosphoribosyltransferase, nicotinate phosphoribosyltransferase domain containing 1, Nicotinate phosphoribosyltransferase domain-containing protein 1, nicotinic acid phosphoribosyltransferase, PP3856
This antibody was developed using a recombinant protein derived from Q6XQN6, with the exact immunogen sequence remaining proprietary.
0.1 mL
Lipid and Metabolism
93100
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL0366
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified