missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NARF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530544
This item is not returnable.
View return policy
Description
Recommended positive controls: A549, H1299, HCT116, MCF-7, mouse muscle. Predicted reactivity: Mouse (92%), Rat (93%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Several proteins have been found to be prenylated and methylated at their carboxyl-terminal ends. Prenylation was initially believed to be important only for membrane attachment. However, another role for prenylation appears to be its importance in protein-protein interactions. The only nuclear proteins known to be prenylated in mammalian cells are prelamin A- and B-type lamins. Prelamin A is farnesylated and carboxymethylated on the cysteine residue of a carboxyl-terminal CaaX motif. This post-translationally modified cysteine residue is removed from prelamin A when it is endoproteolytically processed into mature lamin A. The protein encoded by this gene binds to the prenylated prelamin A carboxyl-terminal tail domain. It may be a component of a prelamin A endoprotease complex. The encoded protein is located in the nucleus, where it partially colocalizes with the nuclear lamina. It shares limited sequence similarity with iron-only bacterial hydrogenases. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene, including one with a novel exon that is generated by RNA editing.
Specifications
| NARF | |
| Polyclonal | |
| Unconjugated | |
| NARF | |
| 4430402O11Rik; IOP2; iron-only hydrogenase-like protein 2; Narf; Nuclear prelamin A recognition factor; prenyl-dependent prelamin A binding protein; RGD1310894 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26502, 67608 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9CYQ7, Q9UHQ1 | |
| NARF | |
| Recombinant protein encompassing a sequence within the center region of human NARF. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction