missing translation for 'onlineSavingsMsg'
Learn More

Naringin Hydrate, TRC

Product Code. 30477212
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477212 25 g -
30476727 1 g -
30464814 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477212 Supplier Toronto Research Chemicals Supplier No. TRCN37898025G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132203-74-8
Molecular Formula C27 H32 O14 . H2 O
Molecular Weight (g/mol) 598.55
Synonym (S)-7-[[2-O-(6-Deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-2,3-dihydro-5-hydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one Hydrate, Naringenin 7-Neohesperidoside Hydrate, Aurantiin Hydrate
IUPAC Name (2S)-7-[(2S,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-5-hydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one;hydrate
SMILES O.C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2Oc3cc(O)c4C(=O)C[C@H](Oc4c3)c5ccc(O)cc5)[C@H](O)[C@H](O)[C@H]1O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C27H32O14.H2O/c1-10-20(32)22(34)24(36)26(37-10)41-25-23(35)21(33)18(9-28)40-27(25)38-13-6-14(30)19-15(31)8-16(39-17(19)7-13)11-2-4-12(29)5-3-11;/h2-7,10,16,18,20-30,32-36H,8-9H2,1H3;1H2/t10-,16-,18+,20-,21+,22+,23-,24+,25+,26-,27+;/m0./s1
Formula Weight 598.1898
Chemical Name or Material Naringin Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.