missing translation for 'onlineSavingsMsg'
Learn More

Naringin, MedChemExpress

Product Code. 16421496
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
10 g
Unit Size:
10g
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16421496 200 mg 200mg
16488878 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16421496 Supplier Medchem Express Supplier No. HYN0153
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Naringin is a major flavanone glycoside obtained from tomatoes, grapefruits, and many other citrus fruits. Naringin exhibits biological properties such as antioxidant, anti-inflammatory, and antiapoptotic activities.

Naringin is a major flavanone glycoside obtained from tomatoes, grapefruits, and many other citrus fruits. Naringin exhibits biological properties such as antioxidant, anti-inflammatory, and antiapoptotic activities.

Specifications

Chemical Name or Material Naringin
CAS 10236-47-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H32O14
Quantity 200 mg
Synonym Naringoside
Solubility Information DMSO : 250 mg/mL (430.64 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.72 mM; ultrasonic and warming and heat to 80°C) ∣Ethanol : 1 mg/mL (1.72 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C1C[C@@H](C2=CC=C(O)C=C2)OC3=CC(O[C@H]4[C@@H]([C@H]([C@@H]([C@@H](CO)O4)O)O)O[C@@]5([H])[C@@H]([C@@H]([C@H]([C@H](C)O5)O)O)O)=CC(O)=C13
Molecular Weight (g/mol) 580.53
Formula Weight 580.53
Percent Purity 98.41%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.