missing translation for 'onlineSavingsMsg'
Learn More

Naringin

Product Code. 15445429
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15445429 25 g 25g
15455429 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15445429 Supplier Thermo Scientific Alfa Aesar Supplier No. J63166.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A citrus bioflavanoid that inhibits cytochrome P-450 monooxygenase activity | CAS: 10236-47-2 | C27H32O14 | 580.54 g/mol

A citrus bioflavanoid that inhibits cytochrome P-450 monooxygenase activityNaringin is used to prepare naringin dihydrochalcone, which is about 300-1800 times sweeter than sugar. It is a citrus bioflavonoid reported to inhibit cytochrome P450 monooxygenase activity. It also acts as an inhibitor of cytochrome P450 enzymes. It is employed as an antioxidant, lipid lowering and anticancer agent. In addition to this, it is used in a study to assess aldosterone production by a human adrenocortical cell line and to improve cognitive deficits of Alzheimer's disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A citrus bioflavanoid that inhibits cytochrome P-450 monooxygenase activityNaringin is used to prepare naringin dihydrochalcone, which is about 300-1800 times sweeter than sugar. It is a citrus bioflavonoid reported to inhibit cytochrome P450 monooxygenase activity. It also acts as an inhibitor of cytochrome P450 enzymes. It is employed as an antioxidant, lipid lowering and anticancer agent. In addition to this, it is used in a study to assess aldosterone production by a human adrenocortical cell line and to improve cognitive deficits of Alzheimer′s disease.

Solubility
Soluble in water, alcohol, acetone and warm acetic acid.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10236-47-2
Molecular Formula C27H32O14
Molecular Weight (g/mol) 580.54
MDL Number MFCD00148888,MFCD00149445,MFCD01461988
InChI Key DFPMSGMNTNDNHN-ZPHOTFPESA-N
Synonym 4',5,7-Trihydroxyflavanone 7-rhamnoglucoside
PubChem CID 74787988
IUPAC Name 7-[(2S,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-5-hydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
SMILES C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2OC2=CC(O)=C3C(=O)C[C@H](OC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@H](O)[C@H]1O

Specifications

Color White to Yellow
Quantity 25 g
Beilstein 102012
Merck Index 14,6425
Solubility Information Soluble in water,alcohol,acetone and warm acetic acid.
Formula Weight 580.53
Physical Form Powder
Chemical Name or Material Naringin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.