missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Narrow Neck Reagent Bottle

Product Code. 10040052
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Height (Metric):
108 mm
136 mm
170 mm
221 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Height (Metric) unitSize
10040052 1000 mL 221 mm Each
10074170 100 mL 108 mm Each
10503631 250 mL 136 mm Each
10184672 500 mL 170 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10040052 Supplier Buerkle™ Supplier No. 03401000
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Universal bottles for storing, packing, taking samples, sampling and transport of liquids.

  • HDPE, transparent
  • PP tamper-evident screw cap, blue
TRUSTED_SUSTAINABILITY

Specifications

Product Type Reagent Bottle
Material HDPE
Includes Polypropylene tamper-evident blue screw cap
Closure Material Polypropylene
Capacity (Metric) 1000 mL
Color Transparent
For Use With (Application) Suitable for Viscous Oils
Height (Metric) 221 mm
Length (Metric) 85 mm
Width (Metric) 85 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.