missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Narrow Neck Reagent Bottle

Product Code. 10074170
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Height (Metric):
108 mm
136 mm
170 mm
221 mm
missing translation for 'unitSize'
Each
This item is not returnable. View return policy

Product Code. 10074170

missing translation for 'mfr': Buerkle™ 03400100

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Universal bottles for storing, packing, taking samples, sampling and transport of liquids.

  • HDPE, transparent
  • PP tamper-evident screw cap, blue
TRUSTED_SUSTAINABILITY

Specifications

Product Type Reagent Bottle
Material HDPE
Includes Polypropylene tamper-evident blue screw cap
Closure Material Polypropylene
Capacity (Metric) 100 mL
Color Transparent
For Use With (Application) Suitable for Viscous Oils
Height (Metric) 108 mm
Length (Metric) 45 mm
Width (Metric) 45 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.