missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Narsoplimab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558704
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Narsoplimab is a human monoclonal antibody targeting mannan-binding lectin-associated serine protease-2 (MASP-2), the effector enzyme of the lectin pathway of the complement system. The lectin pathway is one of the principal pathways of complement and is activated primarily by tissue damage and microbial infection. Importantly, inhibition of MASP-2 does not appear to interfere with the classical complement pathway, a critical component of the acquired immune response to infection. This novel, proprietary pharmaceutical is designed to prevent complement-mediated inflammation and endothelial damage while leaving intact the respective functions of the other pathways of innate immunity.
Specifications
| Narsoplimab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| OMS721 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG4SP |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction