missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAT8 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93546-0.02ml
This item is not returnable.
View return policy
Description
NAT8 Polyclonal antibody specifically detects NAT8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| NAT8 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| Camello-like protein 1, CML1, EC 2.3.1, EC 2.3.1.-, GLAkidney- and liver-specific gene product, Hcml1, N-acetyltransferase 8, N-acetyltransferase 8 (GCN5-related, putative), probable N-acetyltransferase 8, TSC501N-acetyltransferase 8 (camello like), TSC510 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 61-220 of human NAT8 (Q9UHE5). LLALVFSISLFPALWFLAKKPWTEYVDMTLCTDMSDITKSYLSERGSCFWVAESEEKVVGMVGALPVDDPTLREKRLQLFHLFVDSEHRRQGIAKALVRTVLQFARDQGYSEVILDTGTIQLSAMALYQSMGFKKTGQSFFCVWARLVALHTVHFIYHLP | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9027 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction