missing translation for 'onlineSavingsMsg'
Learn More

β-Naththoxyacetic Acid, free acid, 99.1%, MP Biomedicals™

Product Code. 11494981 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494981 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11494981 Supplier MP Biomedicals Supplier No. 0215578925
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

beta-Naphthoxyacetic Acid is a plant growth regulator, which promote the growth of roots on clippings and prevent fruit from falling prematurely. Soluble in alcohol, ehter and acetic acid

TRUSTED_SUSTAINABILITY

Specifications

CAS 120-23-0
Color White
Molecular Formula C12H10O3
MDL Number MFCD00004066
Quantity 25 g
Synonym 2-naphthoxyacetic acid, 2-naphthyloxy acetic acid, bnoa, 2-naphthyloxyacetic acid, 2-naphthalen-2-yloxy acetic acid, noxa, betoxon, 2-noxa, naphthoxyacetic acid, betapal
InChI Key RZCJYMOBWVJQGV-UHFFFAOYSA-N
SMILES C1=CC=C2C=C(C=CC2=C1)OCC(=O)O
IUPAC Name 2-naphthalen-2-yloxyacetic acid
Molecular Weight (g/mol) 202.209
PubChem CID 8422
ChEBI CHEBI:50397
Formula Weight 202.2
Physical Form Crystalline Powder
Chemical Name or Material β-Naththoxyacetic Acid, Free Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.