Learn More
Native Lipoteichoic Acid Mouse anti-Bacteria, Clone: mAb 55, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP1-60146-0.1mg
 View more versions of this product
Product Code. 18749283
  • 548.00 € / 0.10mg
Description
Native Lipoteichoic Acid Monoclonal specifically detects Native Lipoteichoic Acid in Bacteria samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunoassay.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Native Lipoteichoic Acid
Monoclonal
0.2 mg/mL
Western Blot 1:1000, Flow Cytometry 1:250, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay 1.2 ug/ml in PBS
LTA
Microbial mixture of Streptococcus sobrims HG961, HG962, HG970, and HG977
0.1 mg
Primary
Other
Purified
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
mAb 55
Unconjugated
PBS. 1 ml >200 ug/ml) culture medium with a low endotoxin level with 0.02% Sodium Azide
Mouse
Sterile filtered
RUO
The specificity is labled not applicable.
Store at 4C. Do not freeze.
IgG3