missing translation for 'onlineSavingsMsg'
Learn More

Navitoclax, MedChemExpress

Product Code. 16475927
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16475927

Brand: Medchem Express HY10087

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Navitoclax (ABT-263) is a potent and orally active Bcl-2 family protein inhibitor that binds to multiple anti-apoptotic Bcl-2 family proteins, such as Bcl-xL, Bcl-2 and Bcl-w, with a Ki of less than 1 nM.

Navitoclax (ABT-263) is a potent and orally active Bcl-2 family protein inhibitor that binds to multiple anti-apoptotic Bcl-2 family proteins, such as Bcl-xL, Bcl-2 and Bcl-w, with a Ki of less than 1 nM.

Specifications

Chemical Name or Material Navitoclax
CAS 923564-51-6
Color White
Purity Grade Notes Research
Molecular Formula C47H55ClF3N5O6S3
Quantity 5 mg
Synonym ABT-263
Solubility Information DMF : ≥ 100 mg/mL (102.61 mM) ∣DMSO : 75 mg/mL (76.95 mM; Need ultrasonic)
SMILES O=S(C1=CC(S(NC(C2=CC=C(N3CCN(CC4=C(CCC(C)(C4)C)C5=CC=C(Cl)C=C5)CC3)C=C2)=O)(=O)=O)=CC=C1N[C@H](CCN6CCOCC6)CSC7=CC=CC=C7)(C(F)(F)F)=O
Molecular Weight (g/mol) 974.61
Formula Weight 974.61
Percent Purity 96.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.