missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress NBD-Cl

Product Code. 16442409
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442409 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16442409 Supplier MedChemExpress Supplier No. HYD0042
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress NBD-Cl is a nonfluorescent reagent which becomes highly fluorescent after reaction with thiol or amino groups.

Specifications

Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Product Type Dye Reagents
Quantity 1 g
Shipping Condition Room temperature
Solubility DMSO : ≥ 100 mg/mL (501.13 mM) ∣ H2O : 0.67 mg/mL (3.36 mM; Need ultrasonic)
Target HBV
Molecular Weight (g/mol) 199.55 g/mol
CAS 10199-89-0
Synonym NBD chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.