missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NBL1 Monoclonal Antibody (123202)
Rat Monoclonal Antibody
Brand: Invitrogen™ MA561838
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene product is the founding member of the evolutionarily conserved CAN family of proteins, which contain a domain resembling the CTCK motif found in a number of signaling molecules. These proteins are secreted, and act as BMP antagonists by binding to BMPs and preventing them from interacting with their receptors. They may thus play an important role during growth and development. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
| NBL1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q61477 | |
| Nbl1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse DAN, Ala17-Asp178. | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Inhibition Assays, Western Blot | |
| 123202 | |
| Unconjugated | |
| Nbl1 | |
| D1S1733E; D4H1S1733E; DAN; DAN domain family member 1; Dana; DAND1; differential screening-selected gene aberrant in neuroblastoma; N03; NB; Nbl1; neuroblastoma 1, DAN family BMP antagonist; neuroblastoma candidate region, suppression of tumorigenicity 1; neuroblastoma suppressor of tumorigenicity 1; neuroblastoma, suppression of tumorigenicity 1; Neuroblastoma, suppression of tumorigenicity 1 (DNA segment human D1S1733E); NO3; Protein N03; Zinc finger protein DAN | |
| Rat | |
| Protein A/G | |
| RUO | |
| 17965 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction