Learn More
Invitrogen™ NCAPD2 Recombinant Rabbit Monoclonal Antibody (24GB75)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557515
 View more versions of this product
Product Code. 30275282
  • 198.00 € / 20µL
Description
NCAPD2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Regulatory subunit of the condensin complex, a complex required for conversion of interphase chromatin into mitotic-like condense chromosomes. The condensin complex probably introduces positive supercoils into relaxed DNA in the presence of type I topoisomerases and converts nicked DNA into positive knotted forms in the presence of type II topoisomerases. May target the condensin complex to DNA via its C-terminal domain (PubMed:11136719). May promote the resolution of double-strand DNA catenanes (intertwines) between sister chromatids. Condensin-mediated compaction likely increases tension in catenated sister chromatids, providing directionality for type II topoisomerase-mediated strand exchanges toward chromatid decatenation. Required for decatenation of non-centromeric ultrafine DNA bridges during anaphase. Early in neurogenesis, may play an essential role to ensure accurate mitotic chromosome condensation in neuron stem cells, ultimately affecting neuron pool and cortex size (PubMed:27737959). [UniProt].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NCAPD2
Recombinant Monoclonal
Unconjugated
NCAPD2
2810406C15Rik; 2810465G24Rik; Capd2; CAP-D2; chromosome condensation related SMC associated protein 1; chromosome condensation-related SMC-associated protein 1; Chromosome-associated protein D2; CNAP1; Condensin complex subunit 1; hCAP-D2; KIAA0159; mCAP-D2; mKIAA0159; Ncapd2; Non-SMC condensin I complex subunit D2; non-SMC condensin I complex, subunit D2; RGD1562596; XCAP-D2 homolog
Rabbit
Affinity chromatography
RUO
362438, 68298, 9918
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB75
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q15021, Q8K2Z4
NCAPD2
A synthesized peptide derived from human CNAP1 (aa 100-200).
20 μL
Primary
Human, Mouse, Rat