Learn More
NCBP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-41222
 View more versions of this product
Product Code. 18148818
  • 507.00 € / 0.10mg
Description
NCBP1 Polyclonal specifically detects NCBP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NCBP1
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
NCBP1
Antibody was raised against a 17 amino acid peptide near the center of human NCBP1. The immunogen is located within amino acids 420 - 470 of NCBP1. Amino Acid Squence: LSQDPESPKPKFVREV
Affinity Purified
RUO
4686
Human, Mouse, Rat
IgG
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
1 mg/mL
Western Blot 1-2 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence
CBP80nuclear cap binding protein subunit 1, 80kD, NCBP 80 kDa subunit, NCBPMGC2087, nuclear cap binding protein subunit 1, 80kDa, nuclear cap-binding protein subunit 1,80 kDa nuclear cap-binding protein, Sto1
Rabbit
87 kDa
0.1 mg
Primary
At least two isoforms of NCBP1 are known to exist; this antibody will detect both isoforms of NCBP1.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.