missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 € - 326.00 €
Specifications
| Antigen | NCF1 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:500-1:3000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18603312
|
Novus Biologicals
NBP2-98530-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18640175
|
Novus Biologicals
NBP2-98530-50ul |
50 μg |
216.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NCF1 Polyclonal antibody specifically detects NCF1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| NCF1 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology | |
| PBS (pH 7.0) | |
| 653361 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:500-1:3000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| 47 kDa neutrophil oxidase factor, FLJ79451, NADPH oxidase organizer 2, NCF-1, NCF1A, neutrophil cytosol factor 1,47 kDa autosomal chronic granulomatous disease protein, neutrophil cytosolic factor 1, neutrophil cytosolic factor 1 (47kD, chronic granulomatous disease, autosomal1), neutrophil cytosolic factor 1, (chronic granulomatous disease, autosomal 1), Neutrophil NADPH oxidase factor 1, Nox organizer 2, NOXO2NCF-47K, Nox-organizing protein 2, p47phox, SH3 and PX domain-containing protein 1A, SH3PXD1Ap47-phox | |
| Produced in rabbits immunized with E. coli-derived Human NCF1 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title