missing translation for 'onlineSavingsMsg'
Learn More

NCOA7 Monoclonal Antibody (OTI2E12), TrueMAB™, OriGene

Product Code. 17573594
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17573594 30 μL 30µL
17553594 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17573594 Supplier Origene Technologies Supplier No. TA810476S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NCOA7 Monoclonal antibody specifically detects NCOA7 in Human samples. It is validated for Western Blot

Specifications

Antigen NCOA7
Applications Western Blot
Classification Monoclonal
Clone OTI2E12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MED19
Gene Accession No. A0JLT2
Gene Alias 140 kDa estrogen receptor-associated protein; 9030406N13Rik; dJ187J11.3; ERAP140; ESNA1; Estrogen nuclear receptor coactivator 1; estrogen receptor associated protein 140 kDa; Nbla00052; Nbla10993; Ncoa7; NCOA7-AS; Nuclear receptor coactivator 7; putative protein product of Nbla00052; putative protein product of Nbla10993; RGD1566426; TBC/LysM-associated domain containing 4; TLDC4
Host Species Mouse
Immunogen Full length human recombinant protein of human MED19 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219541
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.