missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NCoR2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 30533776
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30533776 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30533776 Supplier Invitrogen™ Supplier No. PA5149286

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

Steroid and thyroid hormones and retinoic acid regulate a complex array of gene expression activity via intracellular receptor transcription factors belonging to the ligand-dependent nuclear receptor superfamily. Adding to the complexity of function of these transcription factors are associated proteins known as coactivators and corepressors which, as their names suggest, enhance or depress transcription activity of the nuclear receptor with which they associate. Silencing mediator of retinoic acid and thyroid hormone receptor (SMRT) and nuclear receptor corepressor (N-CoR) are related transcriptional corepressors which contain two distinct domains capable of interacting with unliganded nuclear receptors to repress their basal transcriptional activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCoR2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene NCOR2
Gene Accession No. Q9Y618
Gene Alias CTG repeat protein 26; CTG26; N-CoR; NCOR2; N-CoR2; nuclear receptor corepressor; nuclear receptor corepressor 2; nuclear receptor co-repressor 2; silencing mediator for retinoid and thyroid hormone receptors; silencing mediator of retinoic acid and thyroid hormone receptor; silencing mediator of retinoid acid and thyroid hormone receptor; SMAP270; SMRT; SMRTe; SMRTE-tau; T3 receptor-associating factor; Thyroid-, retinoic-acid-receptor-associated corepressor; TNRC14; TRAC; TRAC1; TRAC-1
Gene Symbols NCOR2
Host Species Sheep
Immunogen E. coli-derived recombinant human NCOR2, Lys2268-Arg2449.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9612
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.