missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ NCTC-135 Medium
GREENER_CHOICE

Product Code. 11514486 Shop All Gibco Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11514486 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11514486 Supplier Gibco™ Supplier No. 41350026
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

NCTC-135 Medium

NCTC-135 medium is a chemically defined medium that was originally developed at the National Cancer Institute (Bethesda, MD) and formulated for the culture of mouse Strain L sublines, NCTC clone 929 (L929 cells) in a serum-free environment. Gibco™ NCTC-135 medium has also been used to grow many cells, including HeLa, Ehrlich-Lettre Ascites strain E, NCTC clone 1469, NCTC 4206, mouse C3H, BS-C-1, and hybridoma cell lines.

Gibco™ NCTC-135 is a modification of NCTC 109 medium that replaced L-cysteine with L-cystine, which reduced toxicity to some cells. NCTC-135 medium is rich in nucleosides contains several unique components to eliminate the need for serum, including coenzymes, glucosamine, reducing agents and additional vitamins.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the labeled intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ NCTC-135 medium is manufactured at a cGMP compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

NCTC-135 medium contains no proteins, lipids, or growth factors. NCTC-135 medium was designed for use without serum for L929 cells, but typically requires supplementation of a growth factor cocktail or Fetal Bovine Serum (FBS) for other cells. NCTC-135 medium uses a sodium bicarbonate buffer system (2.2 g / L) and therefore requires a 5-10% CO2 environment to maintain physiological pH. Some cell lines might require gradual adaptation to a serum-free medium.
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type NCTC-135 Medium
With Additives Glutamine, Phenol Red, Coenzymes
Without Additives No HEPES
For Use With (Application) Mammalian cell culture
Quantity 500 mL
Certifications/Compliance ISO 13485 standards, FDA
Content And Storage 2 to 8°C
Description NCTC-135 Medium, 500mL, Liquid, L-Glutamine, With L-Glutamine, Phenol Red, Coenzymes, 12 Months Shelf Life, Store at 2 to 8°C, Protect From Light, For Mammalian Cell Culture

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.