missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFA10 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93125-0.1ml
This item is not returnable.
View return policy
Description
NDUFA10 Polyclonal antibody specifically detects NDUFA10 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NDUFA10 | |
| Polyclonal | |
| Western Blot 1:1000-1:4000 | |
| 10 (42kD), CI-42k, CI-42KD, FLJ21923, MGC5103, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 10, 42kDa, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 10, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 36-300 of human NDUFA10 (NP_004535.1). LRYGMWHFLLGDKASKRLTERSRVITVDGNICTGKGKLAKEIAEKLGFKHFPEAGIHYPDSTTGDGKPLATDYNGNCSLEKFYDDPRSNDGNSYRLQSWLYSSRLLQYSDALEHLLTTGQGVVLERSIFSDFVFLEAMYNQGFIRKQCVDHYNEVKSVTICDYLPPHLVIYIDVPVPEVQRRIQKKGDPHEMKITSAYLQDIENAYKKTFLPEMSEKCEVLQYSAREAQDSKKVVEDIEYLKFDKGPWLKQDNRTLYHLRLLVQD | |
| 0.1 mL | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 4705 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction