missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFA9 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94632-0.1ml
This item is not returnable.
View return policy
Description
NDUFA9 Polyclonal antibody specifically detects NDUFA9 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NDUFA9 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CC6, CI39k, CI-39k, CI-39kD, complex I 39kDa subunit, Complex I-39kD, MGC111043, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9 (39kD), NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9, 39kDa, NADH dehydrogenase (ubiquinone) Fe-S protein 2-like (NADH-coenzyme Q reductase), NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 9, mitochondrial, NADH-ubiquinone oxidoreductase 39 kDa subunit, NDUFS2L, SDR22E1, short chain dehydrogenase/reductase family 22E, member 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human NDUFA9 (NP_004993.1). MAAAAQSRVVRVLSMSRSAITAIATSVCHGPPCRQLHHALMPHGKGGRSSVSGIVATVFGATGFLGRYVVNHLGRMGSQVIIPYRCDKYDIMHLRPMGDLGQLLFLEWDARDKDSIRRVVQHSNVVINLIGRDWETKNFDFEDVFVKIPQAIAQLSKEAGVEKFIHVSHLNANIKSSSRYLRNKAVGEKVVRDAFPEAIIVKPSDIFGREDRFLNSFASMHRFGPIPLGSLGWKTVKQPVYVVDVSKGIVNAVKDPDANGKSFAFVGPSR | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 4704 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction