missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFAF3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 572.00 €
Specifications
| Antigen | NDUFAF3 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18414071
|
Novus Biologicals
NBP1-89382-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18221338
|
Novus Biologicals
NBP1-89382 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NDUFAF3 Polyclonal specifically detects NDUFAF3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| NDUFAF3 | |
| Polyclonal | |
| Rabbit | |
| Human, Rat | |
| C3orf60, chromosome 3 open reading frame 60, DKFZp564J0123, DKFZP564J0123,2P1, E3-3, MGC10527, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 3, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 3, nuclear protein E3-3 | |
| NDUFAF3 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 25915 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LLEPRIEIVVVGTGDRTERLQSQVLQAMRQRGIAVEVQDTPNACATFNFLCHEGRVTGAALIPPPGGTSLTSLGQAAQ | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title