missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFB2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94868-0.1ml
This item is not returnable.
View return policy
Description
NDUFB2 Polyclonal antibody specifically detects NDUFB2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| NDUFB2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| CI-AGGGmitochondrial, MGC70788, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 2 (8kD, AGGG), NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 2, 8kDa | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 34-105 of human NDUFB2 (NP_004537.1). AGGGVHIEPRYRQFPQLTRSQVFQSEFFSGLMWFWILWRFWHDSEEVLGHFPYPDPSQWTDEELGIPPDDED | |
| 0.1 mL | |
| Neurodegeneration, Neuroscience | |
| 4708 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction