Learn More
Invitrogen™ NDUFB4 Recombinant Rabbit Monoclonal Antibody (24GB500)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557104
 View more versions of this product
Product Code. 30276611
  • 146.00 € / 20µL
Description
NDUFB4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
The protein encoded by this gene belongs to the complex I NDUFB4 subunit family. Mammalian complex I is composed of 45 different subunits. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NDUFB4
Recombinant Monoclonal
Unconjugated
NDUFB4
0610006N12Rik; 1300010H20Rik; B15; B15 subunit of the NADH: ubiquinone oxidoreductase complex; CI-B15; complex I B15 subunit; complex I-B15; MGC5105; NADH dehydrogenase (ubiquinone) 1 beta subcomplex 4; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 4, 15kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 4; NADH:ubiquinone oxidoreductase B15 subunit; NADH:ubiquinone oxidoreductase subunit B4; NADH-ubiquinone oxidoreductase B15 subunit; Ndufb4; similar to B15 subunit of the NADH: ubiquinone oxidoreductase complex
Rabbit
Affinity chromatography
RUO
288088, 4710, 68194
-20°C
Liquid
Western Blot, Immunocytochemistry
24GB500
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O95168, Q9CQC7
NDUFB4
A synthesized peptide derived from human NDUFB4 (aa 1-129).
20 μL
Primary
Human, Mouse, Rat