missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFS3 Recombinant Rabbit Monoclonal Antibody (2R6F5)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535543
This item is not returnable.
View return policy
Description
Immunogen sequence: WYEREIWDMF GVFFANHPDL RRILTDYGFE GHPFRKDFPL SGYVELRYDD EVKRVVAEPV ELAQEFRKFD LNSPWEAFPV YRQPPESLKL EAGDKKPDAK.
This gene encodes one of the iron-sulfur protein components of mitochondrial NADH:ubiquinone oxidoreductase. Mutations in this gene are associated with Leigh syndrome resulting from mitochondrial complex I deficiency.
Specifications
| NDUFS3 | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75489, Q9DCT2 | |
| NDUFS3 | |
| A synthetic peptide corresponding to a sequence within amino acids 165-264 of human NDUFS3 (O75489). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2R6F5 | |
| Unconjugated | |
| NDUFS3 | |
| 0610010M09Rik; CI-30; CI-30kD; complex I 30kDa subunit; complex I-30kD; hypothetical protein LOC550451; NADH dehydrogenase (ubiquinone); NADH dehydrogenase (ubiquinone) Fe-S protein 3; NADH dehydrogenase (ubiquinone) Fe-S protein 3, (NADH-coenzyme Q reductase); NADH dehydrogenase (ubiquinone) Fe-S protein 3, 30kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 3, mitochondrial; NADH dehydrogenase-ubiquinone 30 kDa subunit; NADH:ubiquinone oxidoreductase core subunit S3; NADH-ubiquinone oxidoreductase 30 kDa subunit; NDUFS3; zgc:112520 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 295923, 4722, 68349 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction