missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFS4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15849-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), or NADH:ubiquinone oxidoreductase, the first multi-subunit enzyme complex of the mitochondrial respiratory chain. Complex I plays a vital role in cellular ATP production, the primary source of energy for many crucial processes in living cells. It removes electrons from NADH and passes them by a series of different protein-coupled redox centers to the electron acceptor ubiquinone. In well-coupled mitochondria, the electron flux leads to ATP generation via the building of a proton gradient across the inner membrane. Complex I is composed of at least 41 subunits, of which 7 are encoded by the mitochondrial genome and the remainder by nuclear genes.Specifications
| NDUFS4 | |
| Polyclonal | |
| Unconjugated | |
| NDUFS4 | |
| AQDQ, CI 18 kDa, CI AQDQ, Complex I 18 kDa, Complex I AQDQ, NDUFS4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17993, 4724, 499529 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O43181, Q5XIF3, Q9CXZ1 | |
| Ndufs4 | |
| NDUFS4 Fusion Protein Ag8633 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction