missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NDUFS6 Recombinant Rabbit Monoclonal Antibody (K01_1U59)
GREENER_CHOICE

Product Code. p-200113720
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830079 100 μL
30827320 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830079 Supplier Invitrogen™ Supplier No. MA563865

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

NDUFS6 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a subunit of the NADH:ubiquinone oxidoreductase, which is the first enzyme complex in the electron transport chain of mitochondria. This complex functions in the transfer of electrons from NADH to the respiratory chain. The subunit encoded by this gene is one of seven subunits in the iron-sulfur protein fraction. Mutations in this gene cause mitochondrial complex I deficiency, a disease that causes a wide variety of clinical disorders, including neonatal disease and adult-onset neurodegenerative disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFS6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_1U59
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene NDUFS6
Gene Accession No. O75380, P52503
Gene Alias BC059730; CI-13kA; CI13KDA; CI-13kD-A; complex I 13kDa subunit A; complex I, mitochondrial respiratory chain, 13-kD subunit; complex I-13kD-A; IP13; Ip13dis; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) Fe-S protein 6; NADH dehydrogenase (ubiquinone) Fe-S protein 6, 13kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 6, mitochondrial; NADH dehydrogenase Fe-S protein 6; NADH ubiquinone oxidoreductase; NADH:ubiquinone oxidoreductase NDUFS6 subunit; NADH:ubiquinone oxidoreductase subunit S6; NADH-ubiquinone oxidoreductase 13 kDa-A subunit; Ndub13; NDUFS6; RATIp13dis; zgc:103774
Gene Symbols NDUFS6
Host Species Rabbit
Immunogen Recombinant protein of human NDUFS6.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 407785, 4726
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.