Learn More
NEDD4 Antibody (S06-7C3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19815-100ul
 View more versions of this product
Product Code. 18648493
  • 645.00 € / 100µL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
NEDD4 Monoclonal antibody specifically detects NEDD4 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NEDD4
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
S06-7C3
Unconjugated
Cell proliferation-inducing gene 53 protein, E3 ubiquitin-protein ligase NEDD4, EC 6.3.2, EC 6.3.2.-, KIAA0093NEDD4-1MGC176705, NEDD-4, Neural precursor cell expressed developmentally down-regulated protein 4, neural precursor cell expressed, developmentally down-regulated 4, receptor-potentiating factor 1, RPF1
A synthetic peptide of human NEDD4 (Uniprot # P46934)
100 μg
Cell Biology, Cell Cycle and Replication, Endocrinology, Immunology, Neuroscience, Signal Transduction
4734
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.