missing translation for 'onlineSavingsMsg'
Learn More

Nefamide, TRC

Product Code. 30482350
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482350 50 mg -
30471928 100 mg -
30481852 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30482350 Supplier Toronto Research Chemicals Supplier No. TRCN38911050MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24833-47-4
Molecular Formula C17H17NO3
Molecular Weight (g/mol) 283.32
Synonym 2-Benzoyl-N-(2-hydroxyethyl)-N-methylbenzamide, o-Benzoyl-N-(2-hydroxyethyl)-N-methylbenzamide
IUPAC Name 2-benzoyl-N-(2-hydroxyethyl)-N-methylbenzamide
SMILES CN(CCO)C(=O)c1ccccc1C(=O)c2ccccc2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H17NO3/c1-18(11-12-19)17(21)15-10-6-5-9-14(15)16(20)13-7-3-2-4-8-13/h2-10,19H,11-12H2,1H3
Formula Weight 283.1208
Chemical Name or Material Nefamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.