missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEFM Monoclonal Antibody (24GB5825)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA557500
This item is not returnable.
View return policy
Description
NEFM Monoclonal Antibody for Western Blot, Flow
Neurofilaments (NFs) are a type of intermediate filament (IF) expressed almost exclusively in neuronal cells, and in those cells most prominently in large axons. NFs, in most vertebrates, are composed of three different polypeptide chains with different molecular weights - neurofilament medium protein (NF-M), high (NF-H) and light protein (NF-L), which share sequence and structural similarity in a coiled-coil core domain, but differ in the length and sequence of their N-termini and more dramatically of their C-termini which in the case of NF-M and NF-H form the flexible extensions that link NFs to each other and to other elements in the cytoplasm. NF-M protein tail-mediated interactions of neurofilaments are critical for size and cytoskeletal architecture of axons, and are mediated, in part, by the highly phosphorylated tail domain of this protein. NF-M phosphorylation and O-GlcNAcylation are regulated reciprocally and affect its translocation and filament formation and function. Antibodies to the various neurofilament subunits are very useful cell type markers since the proteins are among the most abundant of the nervous system, are expressed only in neurons and are biochemically very stable.
Specifications
| NEFM | |
| Monoclonal | |
| Unconjugated | |
| NEFM | |
| 160 kDa neurofilament protein; major neuronal intermediate filament subunit; medium neurofilament subunit; middle molecular weight neurofilament protein NF-M(2); Nef3; Nefm; nef-m; nefm.S; nefm-a; nefm-b; Neurofilament 3; neurofilament 3 (150kDa medium); neurofilament 3, medium; neurofilament M subunit; neurofilament medium; neurofilament medium polypeptide; neurofilament medium S homeolog; neurofilament medium tail domain; neurofilament protein M; Neurofilament protein, middle polypeptide; neurofilament triplet M protein; neurofilament, medium polypeptide; neurofilament, medium polypeptide 150kDa; neurofilament, medium polypeptide S homeolog; neurofilament-3 (150 kD medium); neurofilament-M; neurofilament-M subunit; neuronal intermediate filament; NF160; NF165; NFM; NF-M; NF-M c-terminus; NF-M protein; NMC; XELAEV_18020229mg | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4741 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB5825 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P07197 | |
| NEFM | |
| Recombinant protein of human NEFM (aa 413-773). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction