missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NEK9 Recombinant Rabbit Monoclonal Antibody (K01_1U64)

Product Code. 30829896
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829896 100 μL
30828227 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829896 Supplier Invitrogen™ Supplier No. MA563461
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

NEK9 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

NEK9, a NEK type protein kinase, regulates chromosome alignment and segregation in mitosis. The protein has a N-terminal NIMA-like catalytic domain, a central domain with homology to the guanine nucleotide exchange factor for the GTPase Ran (RCC1), and a C-terminal coiled-coil domain. It is phosphorylated by active p34(Cdc2) and is capable of autophosphorylation and oligomerization. The activated protein binds Nek6 and RCC1, and has been shown to associate with the human homolog of the Drosophila protein Bicaudal D (Bicd2) in vivo and to phosphorylate this coiled-coil protein in vitro. NEK9 expression has been documented in normal human brain. ESTs have been isolated from several tissue libraries, including normal bone, lymph and thymus and cancerous cervix, eye, lymph and placenta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NEK9
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1U64
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene NEK9
Gene Accession No. Q8TD19
Gene Alias APUG; C130021H08Rik; DKFZp434D0935; KIAA1995; LCCS10; MGC138306; MGC16714; NC; NEK 9; NEK8; NEK9; Nercc; Nercc 1 kinase; NERCC1; Nercc1 kinase; never in mitosis A-related kinase 9; NIMA (never in mitosis gene a)- related kinase 9; NIMA (never in mitosis gene a)-related expressed kinase 9; NIMA related kinase 9; NimA-related kinase 8; NIMA-related kinase 9; nimA-related protein kinase 9; Serine/threonine-protein kinase Nek9
Gene Symbols NEK9
Host Species Rabbit
Immunogen A synthetic peptide of human NEK9.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 299204, 91754
Target Species Human, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.