missing translation for 'onlineSavingsMsg'
Learn More

Neodymium @ 1000 μg/g in Hydrocarbon Oil, VHG™

Product Code. 17994266
Change view
Click to view available options
Quantity:
50 g
200 g
Unit Size:
200g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17994266 200 g 200g
17960536 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17994266 Supplier VHG Labs Supplier No. VHGOND1000200G
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Neodymium @ 1000 μg/g in Hydrocarbon Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Neodymium
CAS 7440-00-8
Methods ICP/ICPMS
Analyte or Component Names Neodymium
Shipping Condition Room Temperature
Molecular Formula Nd
Concentration 1000 μg/g
Certifications/Compliance ISO 17034
Synonym Nd
InChI Formula InChI=1S/Nd
SMILES [Nd]
IUPAC Name neodymium
Solvent or Matrix Hydrocarbon Oil
Molecular Weight (g/mol) 144.24
Quantity 200 g
Formula Weight 143.91 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.