missing translation for 'onlineSavingsMsg'
Learn More

Neohesperidin dihydrochalcone hydrate, 98+%

Product Code. 15445219
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15445219 1 g 1g
15455219 5 g 5g
15465219 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15445219 Supplier Thermo Scientific Alfa Aesar Supplier No. J62922.03
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Artificial sweetener derived from citrus | CAS: 20702-77-6 | C28H36O15 | 612.58 g/mol

Neohesperidin dihydrochalcone hydrate, is a flavonoid sweetening agent with potent antioxidant activity. It is antioxidant agent. artificial sweetener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Neohesperidin dihydrochalcone hydrate, is a flavonoid sweetening agent with potent antioxidant activity. It is antioxidant agent. artificial sweetener.

Solubility
Soluble in water to 100 mM (with heating).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20702-77-6
Molecular Formula C28H36O15
Molecular Weight (g/mol) 612.58
MDL Number MFCD00017711
InChI Key ITVGXXMINPYUHD-UHFFFAOYSA-N
Synonym neohesperidin dihydrochalcone, nhdc, neohesperidin dc, neohesperidin dhc, unii-3x476d83qv, ccris 4848, neosperidin dihydrochalcone, neohesperidine dihydrochalcone, 1-4-2-o-6-deoxy-alpha-l-mannopyranosyl-beta-d-glucopyranosyl oxy-2,6-dihydroxyphenyl-3-3-hydroxy-4-methoxyphenyl propan-1-one, 1-4-2-o-6-deoxy-alpha-l-mannopyranosyl-beta-d-glucopyranosyl oxy-2,6-dihydroxyphenyl-3-3-hydroxy-4-methoxyphenyl
PubChem CID 30231
ChEBI CHEBI:83535
IUPAC Name 1-[4-[(2S,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-2,6-dihydroxyphenyl]-3-(3-hydroxy-4-methoxyphenyl)propan-1-one
SMILES COC1=C(O)C=C(CCC(=O)C2=C(O)C=C(OC3OC(CO)C(O)C(O)C3OC3OC(C)C(O)C(O)C3O)C=C2O)C=C1

Specifications

Color Yellow
Quantity 1 g
Merck Index 14,6452
Solubility Information Soluble in water to 100 mM (with heating).
Formula Weight 612.58 (Anhydrous)
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Neohesperidin dihydrochalcone hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.