missing translation for 'onlineSavingsMsg'
Learn More

Neohesperidin Dihydrochalcone, TRC

Product Code. 30134524
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134524 1 g 1g
30003369 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134524 Supplier Toronto Research Chemicals Supplier No. TRCN3899501G
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20702-77-6
Molecular Formula C28 H36 O15
Molecular Weight (g/mol) 612.58
Synonym Neohesperidin-dihydrochalcone, 1-[4-[[2-O-(6-Deoxy-α-l-mannopyranosyl)-β-d-glucopyranosyl]oxy]-2,6-dihydroxyphenyl]-3-(3-hydroxy-4-methoxyphenyl)propan-1-one, 1-Propanone, 1-[4-[[2-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-2,6-dihydroxyphenyl]-3-(3-hydroxy-4-methoxyphenyl)-, Glucopyranoside, 3,5-dihydroxy-4-(3-hydroxy-4-methoxyhydrocinnamoyl)phenyl 2-O-(6-deoxy-α-L-mannopyranosyl)-, β-D- (8CI), Glucopyranoside, 3,5-dihydroxy-4-(3-hydroxy-4-methoxyhydrocinnamoyl)phenyl 2-O-α-L-rhamnopyranosyl- (7CI), 1-[4-[[2-O-(6-Deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-2,6-dihydroxyphenyl]-3-(3-hydroxy-4-methoxyphenyl)-1-propanone, Citrosa, NHDC, Neohesperidin DC, Neohesperidin DHC, Neohesperidin dihydrochalcone, Neohesperidine dihydrochalcone
IUPAC Name 1-[4-[(2S,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-2,6-dihydroxyphenyl]-3-(3-hydroxy-4-methoxyphenyl)propan-1-one
SMILES COc1ccc(CCC(=O)c2c(O)cc(O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)cc2O)cc1O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C28H36O15/c1-11-21(34)23(36)25(38)27(40-11)43-26-24(37)22(35)19(10-29)42-28(26)41-13-8-16(32)20(17(33)9-13)14(30)5-3-12-4-6-18(39-2)15(31)7-12/h4,6-9,11,19,21-29,31-38H,3,5,10H2,1-2H3/t11-,19+,21-,22+,23+,24-,25+,26+,27-,28+/m0/s1
Formula Weight 612.2054
Chemical Name or Material Neohesperidin Dihydrochalcone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.