missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Neomycin ReadyMade™ Solution

Product Code. 30044038
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
10 x 100 mL
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30044038 10 x 100 mL 1 L
30044030 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30044038 Supplier TOKU-E™ Supplier No. N02710x100ML

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Neomycin ReadyMade™ Solution is provided as a sterile, ready-to-use preparation of the broad-spectrum aminoglycoside Neomycin Sulfate in 0.9% NaCl.

  • CAS Number: 1405-10-3
  • Molecular Formula: C23H46N6O13
  • Molecular Weight: 614.64 g/mol
  • Storage Conditions: 2-8°C
  • Form: Liquid
  • Source: Streptomyces fradiae
  • Spectrum: Neomycin is a broad-spectrum antibiotic; however, it is mostly used against Gram-negative bacteria
  • Mechanism of Action: Aminoglycosides target the 30S ribosomal subunit resulting in an inability to read mRNA ultimately producing a faulty or non-existent protein

Spécification

Chemical Name or Material Neomycin Sulfate
Physical Form Liquid
CAS 1405-10-3
pH 5.0 to 7.5
Source Streptomyces fradiae
Packaging Plastic Bottle
Sterile Filtered Yes, 0.22 μm
Sterility Sterile
Color Colorless
Concentration 8.0 to 12.0 mg/mL
Quantity 10 x 100 mL
Purity Grade Notes >95% (HPLC)
Molecular Formula C23H46N6O13
Product Line ReadyMade™ Solutions
Shipping Condition Ambient
Synonym Biosol veterinary, Bykomycin, Fradiomycin sulfate, Lidamycin crème, Neobiotic, Neomix, Otobiotic
IUPAC Name (2R,3S,4R,5R,6R)-5-amino-2-(aminomethyl)-6-[(1R,2R,3S,4R,6S)-4,6-diamino-2-[(2S,3R,4S,5R)-4-[(2R,3R,4R,5S,6S)-3-amino-6-(aminomethyl)-4,5-dihydroxyoxan-2-yl]oxy-3-hydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy-3-hydroxycyclohexyl]oxyoxane-3,4-diol
Molecular Weight (g/mol) 614.64 g/mol
Formula Weight 614.64 amu
Percent Purity >95%
Grade Antibiotic
Concentration or Composition (by Analyte or Components) Neomycin Sulfate ≥1%
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.