missing translation for 'onlineSavingsMsg'
Learn More

Neomycin Sulfate, MP Biomedicals™

Product Code. 17650887 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17650887 5 g 5g
17660887 25 g 25g
17670887 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17650887 Supplier MP Biomedicals Supplier No. 0218061005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Acts against both gram-positive and gram-negative bacteria. Neomycin Sulfate, MP Biomedicals is produced by Streptomyces containing a minimum of 85% neomycin B.

This product acts by binding to the 30S and 50S subunits, causing miscoding and inhibiting initiation and elongation during protein synthesis. Neomycin also blocks voltage-sensitive Ca2+ channels without affecting the Na+/Ca2+ antiporter in neurons.

Applications:

  • As a selection agent for prokaryotic cells transformed using the neo selectable marker gene
  • To study ototoxic side effects of antibiotics and platelet-derived growth factor responses in certain fibroblasts
  • In the extraction of Nuclear Phosphatidylinositol 4,5-Bisphosphate-Interacting Proteins
  • It is recommended for use in cell culture applications at 50mg/L
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-10-3
Molecular Formula C23H48N6O17S
Molecular Weight (g/mol) 712.72
MDL Number MFCD29905465
InChI Key OIXVKQDWLFHVGR-DJUBFIBWNA-N
PubChem CID 124080932
SMILES OS(O)(=O)=O.NCC1OC(O[C@@H]2[C@@H](CO)O[C@@H](OC3C(O)C(N)CC(N)C3OC3OC(CN)C(O)C(O)C3N)[C@@H]2O)C(N)C(O)C1O

Specifications

Physical Form Powder
pH 5 to 7.5
Loss on Drying <8.0%
Quantity 5 g
Solubility Information H2O soluble (50mg/mL)
IUPAC Name 5-amino-2-(aminomethyl)-6-{[(2R,3S,4R,5S)-5-[(3,5-diamino-2-{[3-amino-6-(aminomethyl)-4,5-dihydroxyoxan-2-yl]oxy}-6-hydroxycyclohexyl)oxy]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxy}oxane-3,4-diol; sulfuric acid
Formula Weight 908.9g/mol
Grade USP
compliance-icons
Product Identifier
  • Neomycin Sulfate, USP
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P321-Specific treatment (see on this label).
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Neomycin Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.