missing translation for 'onlineSavingsMsg'
Learn More

Neostigmine bromide

Product Code. 15414709
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15414709 250 mg 250mg
15404709 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15414709 Supplier Thermo Scientific Alfa Aesar Supplier No. J62300.MD
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A reversible acetylcholinesterase inhibitor | CAS: 114-80-7 | C12H19BrN2O2 | 303.20 g/mol

A reversible acetylcholinesterase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114-80-7
Molecular Formula C12H19BrN2O2
Molecular Weight (g/mol) 303.20
MDL Number MFCD00011795
InChI Key LULNWZDBKTWDGK-UHFFFAOYSA-M
Synonym neostigmine bromide, neo-proserin, eustigmin bromide, neoserine bromide, proserine bromide, stigmanol bromide, synstigmin bromide, leostigmine bromide, kirkstigmine bromide, philostigmin bromide
PubChem CID 8246
ChEBI CHEBI:179557
IUPAC Name 3-[(dimethylcarbamoyl)oxy]-N,N,N-trimethylanilinium bromide
SMILES [Br-].CN(C)C(=O)OC1=CC=CC(=C1)[N+](C)(C)C

Specifications

Melting Point 175°C to 177°C
Color White
Quantity 250 mg
UN Number UN2811
Merck Index 14,6464
Formula Weight 303.2
Physical Form Powder
Chemical Name or Material Neostigmine bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.