missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride, 95%

Product Code. 15498076
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498076 250 mg 250mg
15478076 1 g 1g
15488076 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15498076 Supplier Thermo Scientific Alfa Aesar Supplier No. H62993.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13288-57-8 | C15H31ClN2O4 | 338.87 g/mol

Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride is an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride is an pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13288-57-8
Molecular Formula C15H31ClN2O4
Molecular Weight (g/mol) 338.87
MDL Number MFCD00038899
InChI Key TZBPQINFXPIRBX-VAMKTSTMNA-N
Synonym h-lys boc-otbu.hcl, h-lys boc-otbu hcl, ne-boc-l-lysine tert-butyl ester hydrochloride, h-l-lys boc-otbu*hcl, c15h30n2o4.hcl, pubchem18975, lys boc-otbu hcl, h-lys boc-obut hcl, h-lys boc-otbu?hcl, h-lys boc-otbu hydrochloride
PubChem CID 13819885
SMILES Cl.CC(C)(C)OC(=O)NCCCC[C@H](N)C(=O)OC(C)(C)C

Specifications

Color White
Solubility Information Insoluble in water.
IUPAC Name tert-butyl 2-amino-6-{[(tert-butoxy)carbonyl]amino}hexanoate hydrochloride
Quantity 250 mg
Formula Weight 338.87
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Nepsilon-Boc-L-lysine tert-butyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.