missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Boc-Nalpha-Fmoc-L-lysine, 98%

Product Code. 15475418
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475418 25 g 25g
15465418 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475418 Supplier Thermo Scientific Alfa Aesar Supplier No. H59182.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 71989-26-9 | C26H32N2O6 | 468.55 g/mol

It is used to prepare dimeric RGD peptide-paclitaxel conjugate as model for integrin-targeted drug delivery. It is also used to synthesize DOTA-conjugated multivalent cyclic-RGD peptide dendrimers for tumor targeting and imaging use.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to prepare dimeric RGD peptide-paclitaxel conjugate as model for integrin-targeted drug delivery. It is also used to synthesize DOTA-conjugated multivalent cyclic-RGD peptide dendrimers for tumor targeting and imaging use.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71989-26-9
Molecular Formula C26H32N2O6
Molecular Weight (g/mol) 468.55
MDL Number MFCD000371
InChI Key UMRUUWFGLGNQLI-ANBDAQEENA-N
Synonym fmoc-lys boc-oh, fmoc-lys boc, na-fmoc-ne-boc-l-lysine, n-alpha-fmoc-nepsilon-boc-l-lysine, nepsilon-boc-nalpha-fmoc-l-lysine, nepsilon-tert-butoxycarbonyl-nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-lysine, n-fmoc-n'-boc-l-lysine, n-alpha-fmoc-n-epsilon-tert-boc-l-lysine, n, a-fmoc-n, a-boc-l-lysine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid
PubChem CID 2724628
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Specifications

Melting Point 130°C to 135°C
Quantity 25 g
Beilstein 4217767
Solubility Information Slightly soluble in water.
Optical Rotation −12° (c=1 in DMF)
Formula Weight 468.54
Percent Purity 98%
Chemical Name or Material Nepsilon-Boc-Nalpha-Fmoc-L-lysine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.