missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Trifluoroacetyl-L-lysine, 97%

Product Code. 15497727
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15497727 5 g 5g
15407737 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497727 Supplier Thermo Scientific Alfa Aesar Supplier No. H37051.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 10009-20-8 | C8H13F3N2O3 | 242.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10009-20-8
Molecular Formula C8H13F3N2O3
Molecular Weight (g/mol) 242.20
MDL Number MFCD00037223
InChI Key PZZHRSVBHRVIMI-MLHKIVSYNA-N
Synonym h-lys tfa-oh, n-6-trifluoroacetyl-l-lysine, n6-trifluoroacetyl-l-lysine, n6-trifluoroaccety-l-lysine, epsilon-tfa-lysine, nepsilon-trifluoroacetyl-l-lysine, l-lysine, n6-trifluoroacetyl, n 6-trifluoroacetyl-l-lysine, n-, a-trifluoroacetyl-l-lysine
PubChem CID 7009573
ChEBI CHEBI:61064
SMILES N[C@@H](CCCCNC(=O)C(F)(F)F)C(O)=O

Specifications

Melting Point ∽253°C (decomposition)
IUPAC Name (2S)-2-amino-6-[(2,2,2-trifluoroacetyl)amino]hexanoic acid
Quantity 5 g
Formula Weight 242.2
Optical Rotation +19° (c=1 in 1M HCl)
Percent Purity 97%
Chemical Name or Material Nepsilon-Trifluoroacetyl-L-lysine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.