Learn More
Thermo Scientific™ NERL™ High Purity Water
Improve control in critical diagnostic procedures by using Thermo Scientific™ NERL™ High Purity Water in place of deionized or distilled water.
93.00 €
Specifications
| pH | 5 to 7 |
|---|---|
| Grade | NERL |
Alert:
- For in vitro diagnostic and laboratory use only
- Not for infusion, injection or irrigation
Description
Improve control in critical diagnostic procedures by using Thermo Scientific™ NERL™ High Purity Water in place of deionized or distilled water. This product meets CAP/CLSI specifications for Clinical Laboratory Reagent Grade Water (CLRW) at time of manufacture.
Typically prepared at 18 megohm/cm specific resistance using reverse osmosis, mixed bed deionization, activated carbon filtration and final filtration to 0.2 micron, NERL high purity water is essentially free from organic and inorganic particulate and soluble contaminants.
Contains no preservatives
Ordering Information:
Certificate of analysis available for each case
Specifications
| 5 to 7 | |
| NERL |
Safety and Handling
Recommended Storage : 15° to 30°C
General Purpose - For in vitro diagnostic or laboratory use only. Not for infusion, injection or irrigation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.