Learn More
Nestin Antibody (196908), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB1259MFV450
Product Code. 18519416
  • 719.00 € / 0.10mL
Description
Nestin Monoclonal antibody specifically detects Nestin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-reported
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nestin
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Mouse
0.1 mL
Angiogenesis, Cancer, Cancer Stem Cells, Cell Biology, Cellular Markers, Extracellular Matrix, Neuronal Cell Markers, Neuroscience, Stem Cell Markers
10763
Store at 4C in the dark.
Flow Cytometry, Immunocytochemistry, CyTOF
196908
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-reported
FLJ21841, nestin
NS0 mouse myeloma cell line transfected with human Nestin
RUO
Primary
Human
IgG1