Learn More
Nestin Antibody (CL0197) [Alexa Fluor™ 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59775AF350
Product Code. 30302731
  • 759.00 € / 0.10mL
Description
Nestin Monoclonal antibody specifically detects Nestin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nestin
Monoclonal
Alexa Fluor 350
FLJ21841, nestin
This antibody was developed using a recombinant protein derived from P48681, with the exact immunogen sequence remaining proprietary.
0.1 mL
Angiogenesis, Cancer, Cancer Stem Cells, Cell Biology, Cellular Markers, Extracellular Matrix, Neuronal Cell Markers, Neuroscience, Stem Cell Markers
10763
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence
CL0197
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified